1,6-Diphenoxy-2,4-hexadiin

Lieferant: Molekula

Synonyme: 1,6-Diphenoxyhexa-2,4-diyne

M79271366
MOLEM79271366EA 0 EUR
MOLEM79271366
1,6-Diphenoxy-2,4-hexadiin
1,6-Diphenoxy-2,4-hexadiin
Formel: C₁₈H₁₄O₂
Molecular Weight: 262,31 g/mol
MDL: MFCD00041712
CAS-Nummer: 30980-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR