Tetradecyltrimethylammoniumbromid

Lieferant: Molekula
Gefahr

M79329880
MOLEM79329880EA 0 EUR
MOLEM79329880
Tetradecyltrimethylammoniumbromid
Tetradecyltrimethylammoniumbromid
Formel: C₁₇H₃₈BrN
Schmelzpunkt: 245…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011770
CAS-Nummer: 1119-97-7
EINECS: 214-291-9
UN: 3261
ADR: 8,II
Merck Index: 12,06419

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR