(4-Nitrophenyl)-α-D-glucopyranosid

Lieferant: Molekula

Synonyme: 4-Nitrophenyl α-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl a-D-glucopyranoside

M79358495
MOLEM79358495EA 0 EUR
MOLEM79358495
(4-Nitrophenyl)-α-D-glucopyranosid
(4-Nitrophenyl)-α-D-glucopyranosid
Formel: C₁₂H₁₅NO₈
Molecular Weight: 301,25 g/mol
MDL: MFCD00064088
CAS-Nummer: 3767-28-0
EINECS: 223-189-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR