3-Brom-p-toluidin
Lieferant: Molekula
Synonyme:
3-Brom-4-methylanilin, 4-Amino-2-bromtoluol
M79676383
MOLEM79676383EA
0
EUR
MOLEM79676383
3-Brom-p-toluidin
3-Brom-p-toluidin
Formel:
C₇H₈BrN Molecular Weight: 186,05 g/mol Siedepunkt: 254…257 °C (1013 hPa) Schmelzpunkt: 25…26 °C |
MDL:
MFCD00134176 CAS-Nummer: 7745-91-7 EINECS: 231-807-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...