2-(Trifluormethyl)phenylhydrazin

Lieferant: Molekula

Synonyme: 1-(2-(Trifluormethyl)phenyl)hydrazin, 2-Hydrazinobenzotrifluorid

M79900845
MOLEM79900845EA 0 EUR
MOLEM79900845
2-(Trifluormethyl)phenylhydrazin
2-(Trifluormethyl)phenylhydrazin
Formel: C₇H₇F₃N₂
Molecular Weight: 176,14 g/mol
MDL: MFCD00052687
CAS-Nummer: 365-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR