DL-Norvalin

Lieferant: Molekula

Synonyme: (±)-2-Aminovaleriansäure, (±)-Norvalin, H-DL-Nva-OH, (±)-2-Aminopentansäure

M80205512
MOLEM80205512EA 0 EUR
MOLEM80205512
DL-Norvalin
DL-Norvalin
Formel: C₅H₁₁NO₂
Molecular Weight: 117,15 g/mol
Siedepunkt: 223 °C (1013 hPa)
Schmelzpunkt: > 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064420
CAS-Nummer: 760-78-1
EINECS: 212-082-7
Merck Index: 12,06813

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR