Methotrexat Hydrat
Lieferant: Molekula
Synonyme:
L-Amethopterin hydrate, Methylaminopterin hydrate, MTX hydrate, (4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoyl)-L-glutamic acid hydrate(1:x), Antifolan hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, (+)-Amethopterin hydrate, L(+)-Amethopterin hydrate, 4-Amino-10-methylfolic acid hydrate
M80419974
MOLEM80419974EA
0
EUR
MOLEM80419974
Methotrexat Hydrat
Methotrexat Hydrat
Formel:
C₂₀H₂₂N₈O₅·xH₂O Molecular Weight: 454,44 g/mol (anhy) |
MDL:
MFCD00150847 CAS-Nummer: 133073-73-1 EINECS: 200-413-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...