p-Tolylhydraziniumchlorid

Lieferant: Molekula

Synonyme: 4-Methylphenylhydrazin Hydrochlorid, 1-(4-Methylphenyl)hydrazin Hydrochlorid, 4-Methylphenylhydrazinhydrochlorid, 4-Tolylhydrazin Hydrochlorid, 4-Tolylhydrazinhydrochlorid, 4-Methylphenylhydraziniumchlorid, 1-(4-Tolyl)hydrazin Hydrochlorid

M80553789
MOLEM80553789EA 0 EUR
MOLEM80553789
p-Tolylhydraziniumchlorid
p-Tolylhydraziniumchlorid
Formel: C₇H₁₁ClN₂
Molecular Weight: 158,63 g/mol
MDL: MFCD00012940
CAS-Nummer: 637-60-5
EINECS: 211-295-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR