8-Bromchinolin

Lieferant: Molekula
Achtung

M80855770
MOLEM80855770EA 0 EUR
MOLEM80855770
8-Bromchinolin
8-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00191859
CAS-Nummer: 16567-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR