5-Methylthiophen-2-carbaldehyd
Lieferant: Molekula
Synonyme:
5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde
M80973270
MOLEM80973270EA
0
EUR
MOLEM80973270
5-Methylthiophen-2-carbaldehyd
5-Methylthiophen-2-carbaldehyd
Formel:
C₆H₆OS Molecular Weight: 126,18 g/mol |
MDL:
MFCD00005434 CAS-Nummer: 13679-70-4 EINECS: 237-178-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...