(±)-Indolin-2-carbonsäure

Lieferant: Molekula

Synonyme: Indoline-2-carboxylic acid

M81083934
MOLEM81083934EA 0 EUR
MOLEM81083934
(±)-Indolin-2-carbonsäure
(±)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
MDL: MFCD00010635
CAS-Nummer: 78348-24-0
EINECS: 278-899-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR