Adenosin-5'-monophosphorsäure (AMP) Monohydrat

Lieferant: Molekula

Synonyme: A-5′-P, AMP, Adenosine-5'-monophosphoric acid monohydrate, 5′-AMP, 5′-Adenylic acid

M81117744
MOLEM81117744EA 0 EUR
MOLEM81117744
Adenosin-5'-monophosphorsäure (AMP) Monohydrat
Adenosin-5'-monophosphorsäure (AMP) Monohydrat
Formel: C₁₀H₁₄N₅O₇P·1H₂O
Molecular Weight: 365.24 g/mol
MDL: MFCD00149360
CAS-Nummer: 18422-05-4
EINECS: 200-500-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR