5-Methyl-2-thiophenboronsäure

Lieferant: Molekula

Synonyme: 5-Methylthiophene-2-boronic acid, (5-Methylthiophen-2-yl)boronic acid

M81164883
MOLEM81164883EA 0 EUR
MOLEM81164883
5-Methyl-2-thiophenboronsäure
5-Methyl-2-thiopheneboronic acid
Formel: C₅H₇BO₂S
Molecular Weight: 141,99 g/mol
MDL: MFCD01318166
CAS-Nummer: 162607-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR