1,3-Bis(trimethylsilyl)harnstoff

Lieferant: Molekula

Synonyme: N,N'-Bis(trimethylsilyl)urea

M81616581
MOLEM81616581EA 0 EUR
MOLEM81616581
1,3-Bis(trimethylsilyl)harnstoff
1,3-Bis(trimethylsilyl)harnstoff
Formel: C₇H₂₀N₂OSi₂
MDL: MFCD00008260
CAS-Nummer: 18297-63-7
EINECS: 242-177-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR