5,6-Dimethoxyindol

Lieferant: Molekula

M82103323
MOLEM82103323EA 0 EUR
MOLEM82103323
5,6-Dimethoxyindol
5,6-Dimethoxyindol
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
MDL: MFCD00005675
CAS-Nummer: 14430-23-0
EINECS: 238-402-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR