5-Hydroxyindol
Lieferant: Molekula
Synonyme:
1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indol
M82184513
MOLEM82184513EA
0
EUR
MOLEM82184513
5-Hydroxyindol
5-Hydroxyindol
Formel:
C₈H₇NO Molecular Weight: 133,15 g/mol Schmelzpunkt: 107…108 °C |
MDL:
MFCD00005677 CAS-Nummer: 1953-54-4 EINECS: 217-782-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...