D-Norvalin
Lieferant: Molekula
Synonyme:
(R)-(-)-2-Aminopentansäure, (R)-(-)-Norvalin, (R)-(-)-2-Aminovaleriansäure, H-D-Nva-OH
M82229985
MOLEM82229985EA
0
EUR
MOLEM82229985
D-Norvalin
D-Norvalin
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol |
MDL:
MFCD00008097 CAS-Nummer: 2013-12-9 EINECS: 217-936-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...