o-Kresolphthalein-Komplexon

Lieferant: Molekula

Synonyme: Phthaleinpurpur

M82452467
MOLEM82452467EA 0 EUR
MOLEM82452467
o-Kresolphthalein-Komplexon
o-Kresolphthalein-Komplexon
Formel: C₃₂H₃₂N₂O₁₂
Molecular Weight: 636,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005911
CAS-Nummer: 2411-89-4
EINECS: 219-318-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR