D(-)-Serin

Lieferant: Molekula

Synonyme: H-D-Ser-OH, (R)-(-)-2-Amino-3-hydroxypropionsäure, (R)-(-)-Serin

M82519233
MOLEM82519233EA 0 EUR
MOLEM82519233
D(-)-Serin
D(-)-Serin
Formel: C₃H₇NO₃
Molecular Weight: 105,09 g/mol
Schmelzpunkt: 235 °C
MDL: MFCD00004269
CAS-Nummer: 312-84-5
EINECS: 206-229-4
Merck Index: 12,08604

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR