Octyl-β-D-thioglucopyranosid

Lieferant: Molekula

Synonyme: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside

M82602895
MOLEM82602895EA 0 EUR
MOLEM82602895
Octyl-β-D-thioglucopyranosid
Octyl-β-D-thioglucopyranosid
Formel: C₁₄H₂₈O₅S
Molecular Weight: 308,44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012189
CAS-Nummer: 85618-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR