D(+)-Cellobioseoctaacetat

Lieferant: Molekula

M82788502
MOLEM82788502EA 0 EUR
MOLEM82788502
D(+)-Cellobioseoctaacetat
D(+)-Cellobioseoctaacetat
Formel: C₂₈H₃₈O₁₉
Molecular Weight: 678,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009600
CAS-Nummer: 5346-90-7
EINECS: 226-304-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR