1,4-Bis(diphenylphosphino)butan
Lieferant: Molekula
Synonyme:
Butan-1,3-diylbis(diphenylphosphin), Butylenbis(diphenylphosphin), Bis(1,2-diphenylphosphino)butan
M82971615
MOLEM82971615EA
0
EUR
MOLEM82971615
1,4-Bis(diphenylphosphino)butan
1,4-Bis(diphenylphosphino)butan
Formel:
C₂₈H₂₈P₂ Molecular Weight: 426,48 g/mol Schmelzpunkt: 195 °C Dichte: 1,23 g/cm³ (20 °C) |
MDL:
MFCD00003051 CAS-Nummer: 7688-25-7 EINECS: 231-698-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...