Diethyl(2-cyanethyl)malonat

Lieferant: Molekula

Synonyme: Diethyl 2-(2-cyanoethyl)malonate

M83241955
MOLEM83241955EA 0 EUR
MOLEM83241955
Diethyl(2-cyanethyl)malonat
Diethyl(2-cyanethyl)malonat
Formel: C₁₀H₁₅NO₄
Molecular Weight: 213,23 g/mol
MDL: MFCD00001966
CAS-Nummer: 17216-62-5
EINECS: 241-260-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR