2-(3-Fluorphenyl)ethanol

Lieferant: Molekula

M83247340
MOLEM83247340EA 0 EUR
MOLEM83247340
2-(3-Fluorphenyl)ethanol
2-(3-Fluorphenyl)ethanol
Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
Siedepunkt: 70…71 °C (2 mmHg)
Dichte: 1,12 g/cm³ (20 °C)
MDL: MFCD00045998
CAS-Nummer: 52059-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR