2,4,5-Trimethoxybenzaldehyd
Lieferant: Molekula
M83334197
MOLEM83334197EA
0
EUR
MOLEM83334197
2,4,5-Trimethoxybenzaldehyd
2,4,5-Trimethoxybenzaldehyde
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,2 g/mol Siedepunkt: 335 °C (1013 hPa) Schmelzpunkt: 112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003312 CAS-Nummer: 4460-86-0 EINECS: 224-713-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...