Triethylammoniumbromid

Lieferant: Molekula

Synonyme: Triethylamin Hydrobromid

M83528342
MOLEM83528342EA 0 EUR
MOLEM83528342
Triethylammoniumbromid
Triethylammonium bromide
Formel: C₆H₁₅N·HBr
Molecular Weight: 182,1 g/mol
MDL: MFCD00054287
CAS-Nummer: 636-70-4
EINECS: 211-263-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR