3-(Trifluormethyl)phenylhydrazin

Lieferant: Molekula

M83582032
MOLEM83582032EA 0 EUR
MOLEM83582032
3-(Trifluormethyl)phenylhydrazin
3-(Trifluormethyl)phenylhydrazin
Formel: C₇H₇F₃N₂
MDL: MFCD00025093
CAS-Nummer: 368-78-5
EINECS: 206-713-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR