Bis(cyclopentadienyl)titandichlorid

Lieferant: Molekula

Synonyme: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride

M83725447
MOLEM83725447EA 0 EUR
MOLEM83725447
Bis(cyclopentadienyl)titandichlorid
Bis(cyclopentadienyl)titandichlorid
Formel: C₁₀H₁₀Cl₂Ti
Molecular Weight: 248,97 g/mol
MDL: MFCD00003723
CAS-Nummer: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR