2-Chlor-6-(trifluormethyl)pyridin

Lieferant: Molekula

M83914845
MOLEM83914845EA 0 EUR
MOLEM83914845
2-Chlor-6-(trifluormethyl)pyridin
2-Chloro-6-(trifluoromethyl)pyridine
Formel: C₆H₃ClF₃N
Molecular Weight: 181,54 g/mol
MDL: MFCD00042226
CAS-Nummer: 39890-95-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR