1,3,5-Triazin

Lieferant: Molekula

Synonyme: s-Triazin

M83926748
MOLEM83926748EA 0 EUR
MOLEM83926748
1,3,5-Triazin
1,3,5-Triazine
Formel: C₃H₃N₃
Molecular Weight: 81,08 g/mol
MDL: MFCD00006044
CAS-Nummer: 290-87-9
EINECS: 206-028-1
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR