trans-2,3-Dibrom-2-buten-1,4-diol

Lieferant: Molekula

M83988571
MOLEM83988571EA 0 EUR
MOLEM83988571
trans-2,3-Dibrom-2-buten-1,4-diol
trans-2,3-Dibrom-2-buten-1,4-diol
Formel: C₄H₆Br₂O₂
MDL: MFCD00004698
CAS-Nummer: 21285-46-1
EINECS: 244-313-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR