Methylcarbazat
Lieferant: Molekula
Synonyme:
Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate
M84177508
MOLEM84177508EA
0
EUR
MOLEM84177508
Methylcarbazat
Methylcarbazat
Formel:
C₂H₆N₂O₂ Molecular Weight: 90,08 g/mol |
MDL:
MFCD00007594 CAS-Nummer: 6294-89-9 EINECS: 228-560-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...