Natrium-3-nitrobenzolsulfonat

Lieferant: Molekula

M84233913
MOLEM84233913EA 160 EUR
MOLEM84233913
Natrium-3-nitrobenzolsulfonat
Natrium-3-nitrobenzolsulfonat
Formel: C₆H₄NNaO₅S
MDL: MFCD00007490
CAS-Nummer: 127-68-4
EINECS: 204-857-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR