O-Methylhydroxylamin Hydrochlorid

Lieferant: Molekula

Synonyme: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride

M84243443
MOLEM84243443EA 0 EUR
MOLEM84243443
O-Methylhydroxylamin Hydrochlorid
O-Methylhydroxylamin Hydrochlorid
Formel: CH₆ClNO
Molecular Weight: 83.515 g/mol
MDL: MFCD00012951
CAS-Nummer: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR