Dimethyl-3-oxoglutarat

Lieferant: Molekula

Synonyme: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

M84420468
MOLEM84420468EA 0 EUR
MOLEM84420468
Dimethyl-3-oxoglutarat
Dimethyl-3-oxoglutarat
Formel: C₇H₁₀O₅
Molecular Weight: 174,15 g/mol
MDL: MFCD00008462
CAS-Nummer: 1830-54-2
EINECS: 217-385-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR