Furan-2-boronsäure

Lieferant: Molekula
Achtung

Synonyme: Furan-2-ylboronic acid

M84472096
MOLEM84472096EA 0 EUR
MOLEM84472096
Furan-2-boronsäure
Furan-2-boronsäure
Formel: C₄H₅BO₃
Molecular Weight: 111,89 g/mol
MDL: MFCD00799544
CAS-Nummer: 13331-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR