Acriflavin Hydrochlorid

Lieferant: Molekula

Synonyme: 3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine

M84900728
MOLEM84900728EA 0 EUR
MOLEM84900728
Acriflavin Hydrochlorid
Acriflavin Hydrochlorid
Formel: C₂₇H₂₈Cl₄N₆
Molecular Weight: 578,36 g/mol
MDL: MFCD00069039
CAS-Nummer: 8063-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR