5-Amino-2,4,6-triiodisophthalsäure

Lieferant: Molekula

Synonyme: 5-Amino-2,4,6-triiodoisophthalic Acid

M85093613
MOLEM85093613EA 0 EUR
MOLEM85093613
5-Amino-2,4,6-triiodisophthalsäure
5-Amino-2,4,6-triiodisophthalsäure
Formel: C₈H₄I₃NO₄
Molecular Weight: 558,84 g/mol
MDL: MFCD00190167
CAS-Nummer: 35453-19-1
EINECS: 252-575-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR