2,2'-Bipyridin-4,4'-dicarbonsäure

Lieferant: Molekula

Synonyme: 4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid

M85271529
MOLEM85271529EA 0 EUR
MOLEM85271529
2,2'-Bipyridin-4,4'-dicarbonsäure
2,2'-Bipyridin-4,4'-dicarbonsäure
Formel: C₁₂H₈N₂O₄
Molecular Weight: 244.20 g/mol
MDL: MFCD00015430
CAS-Nummer: 6813-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR