Tryptamin Hydrochlorid

Lieferant: Molekula

M85276982
MOLEM85276982EA 0 EUR
MOLEM85276982
Tryptamin Hydrochlorid
Tryptamin Hydrochlorid
Formel: C₁₀H₁₃ClN₂
Molecular Weight: 196,68 g/mol
MDL: MFCD00012682
CAS-Nummer: 343-94-2
EINECS: 206-446-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR