D-Phenylalaninol

Lieferant: Molekula

M85573823
MOLEM85573823EA 0 EUR
MOLEM85573823
D-Phenylalaninol
D-Phenylalaninol
Formel: C₉H₁₃NO
MDL: MFCD00064298
CAS-Nummer: 5267-64-1
EINECS: 226-086-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR