2'-Deoxyuridin

Lieferant: Molekula

M85585786
MOLEM85585786EA 0 EUR
MOLEM85585786
2'-Deoxyuridin
2'-Deoxyuridin
Formel: C₉H₁₂N₂O₅
Molecular Weight: 228,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006527
CAS-Nummer: 951-78-0
EINECS: 213-455-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR