5-(2-Bromphenyl)-1H-tetrazol
Lieferant: Molekula
Synonyme:
5-(2-Bromophenyl)tetrazole
M85598531
MOLEM85598531EA
0
EUR
MOLEM85598531
5-(2-Bromphenyl)-1H-tetrazol
5-(2-Bromphenyl)-1H-tetrazol
Formel:
C₇H₅BrN₄ Molecular Weight: 225.05 g/mol Schmelzpunkt: 183…184 °C |
MDL:
MFCD00151799 CAS-Nummer: 73096-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...