Ethylindol-3-ylacetat

Lieferant: Molekula

Synonyme: 3-Indolylessigsäureethylester, Indol-3-ylessigsäureethylester, Ethylindol-3-acetat, Indol-3-essigsäureethylester

M85598997
MOLEM85598997EA 0 EUR
MOLEM85598997
Ethylindol-3-ylacetat
Ethylindol-3-ylacetat
Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005635
CAS-Nummer: 778-82-5
EINECS: 212-296-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR