2-(2-Bromethyl)-1,3-dioxolan

Lieferant: Molekula

M85647981
MOLEM85647981EA 0 EUR
MOLEM85647981
2-(2-Bromethyl)-1,3-dioxolan
2-(2-Bromethyl)-1,3-dioxolan
Formel: C₅H₉BrO₂
Molecular Weight: 181,03 g/mol
MDL: MFCD00003216
CAS-Nummer: 18742-02-4
EINECS: 242-551-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR