1,1,3,3-Tetramethyl-2-thioharnstoff
Lieferant: Molekula
Synonyme:
Tetramethylthioharnstoff, Tetramethyl-2-thioharnstoff, N,N,N',N'-Tetramethylthioharnstoff
M85812929
MOLEM85812929EA
0
EUR
MOLEM85812929
1,1,3,3-Tetramethyl-2-thioharnstoff
1,1,3,3-Tetramethyl-2-thioharnstoff
Formel:
C₅H₁₂N₂S Molecular Weight: 132,23 g/mol Siedepunkt: 245 °C (1013 hPa) Schmelzpunkt: 74…76 °C |
MDL:
MFCD00008324 CAS-Nummer: 2782-91-4 EINECS: 220-488-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...