Platin(IV)oxid
Lieferant: Molekula
|
Achtung
|
Synonyme:
Platindioxid, Adams Katalysator
M86173878
MOLEM86173878EA
0
EUR
MOLEM86173878
Platin(IV)oxid
Platin(IV)oxid
Formel:
O₂Pt Molecular Weight: 227,08 g/mol Schmelzpunkt: 450 °C Dichte: 11,8 g/cm³ (20 °C) |
MDL:
MFCD00011184 CAS-Nummer: 1314-15-4 EINECS: 215-223-0 UN: 1479 ADR: 5.1,II Merck Index: 12,07686 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...