Tetrabutylammoniumtrifluormethansulfonat

Lieferant: Molekula

M86203465
MOLEM86203465EA 0 EUR
MOLEM86203465
Tetrabutylammoniumtrifluormethansulfonat
Tetrabutylammonium trifluoromethanesulphonate
Formel: C₁₇H₃₆F₃NO₃S
Molecular Weight: 391,54 g/mol
MDL: MFCD00042585
CAS-Nummer: 35895-70-6
EINECS: 252-783-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR