1,2,3,4-Tetrahydroisochinolin

Lieferant: Molekula

M86277908
MOLEM86277908EA 0 EUR
MOLEM86277908
1,2,3,4-Tetrahydroisochinolin
1,2,3,4-Tetrahydroisochinolin
Formel: C₉H₁₁N
Molecular Weight: 133,19 g/mol
MDL: MFCD00006896
CAS-Nummer: 91-21-4
EINECS: 202-050-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR