Dimethyl-5-aminoisophthalat
Lieferant: Molekula
Synonyme:
5-Amino-1,3-benzoldicarbonsäuredimethylester, 5-Aminoisophthalsäuredimethylester
M86296751
MOLEM86296751EA
0
EUR
MOLEM86296751
Dimethyl-5-aminoisophthalat
Dimethyl-5-aminoisophthalat
Formel:
C₁₀H₁₁NO₄ Molecular Weight: 209,2 g/mol |
MDL:
MFCD00008435 CAS-Nummer: 99-27-4 EINECS: 202-743-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...